Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC9A7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567384
Description
Immunogen sequence: SWLNIRVGVEE PSEEDQNEHH WQYFRVGVDP DQDPPPNNDS FQVLQGDGPD SARGNRTKQE SAWIFRLWY Highest antigen sequence identity to the following orthologs - mouse 90%, rat 91%.
Na+/H+ exchangers (NHE) of mammalian cells are plasma membrane intrinsic proteins mediating exchange of N+ and H+ ions in various tissues. The NHE catalyzes the electroneural transport of extracellular Na+ for intracellular H+. They play a major role in regulation of intracellular pH (pHi) in addition to trans-cellular absorption of Na+, cell volume regulation and possibly in cell proliferation. These primary functions of the Na+/H+ exchanger have been related to many pathophysiological states, include hypertension, organ growth and hypertrophy, regression of cancer and renal intestinal disorders. At least 7 NHE isoforms (NHE1-7) have been cloned so far. They are all similar in their primary structure and predicted to have 10-12 transmembrane domains. The C-terminal domain of NHEs are predicted to be intracellular. NHE7 (human 725 aa, chromosome Xp11.4) is ubiquitously expressed, and predominantly localizes to the trans-golgi network. NHE7 mediates the influx of Na+ or K+ in exchange for H+. It is ~70% related to NHE6 but relatively less (~25%) homologous with other NHEs.
Specifications
SLC9A7 | |
Polyclonal | |
Unconjugated | |
SLC9A7 | |
A530087D17Rik; hypothetical protein LOC554532; Na(+)/H(+) exchanger 7; Nhe7; NHE-7; nonselective sodium potassium/proton exchanger; SLC9A6; Slc9a7; Sodium/hydrogen exchanger 7; solute carrier family 9 (sodium/hydrogen exchanger), isoform 7; solute carrier family 9 (sodium/hydrogen exchanger), member 7; solute carrier family 9 member 7; solute carrier family 9 member A7; solute carrier family 9, subfamily A (NHE7, cation proton antiporter 7), member 7; wu:fk03b09; zgc:101088; zgc:113878 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84679 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96T83 | |
SLC9A7 | |
Recombinant Human SLC9A7. Recombinant protein control fragment (Product #RP-108044). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction