Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC9A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106748
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106748 Supplier Invitrogen™ Supplier No. PA5106748
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC9A7.

Na+/H+ exchangers (NHE) of mammalian cells are plasma membrane intrinsic proteins mediating exchange of N+ and H+ ions in various tissues. The NHE catalyzes the electroneural transport of extracellular Na+ for intracellular H+. They play a major role in regulation of intracellular pH (pHi) in addition to trans-cellular absorption of Na+, cell volume regulation and possibly in cell proliferation. These primary functions of the Na+/H+ exchanger have been related to many pathophysiological states, include hypertension, organ growth and hypertrophy, regression of cancer and renal intestinal disorders. At least 7 NHE isoforms (NHE1-7) have been cloned so far. They are all similar in their primary structure and predicted to have 10-12 transmembrane domains. The C-terminal domain of NHEs are predicted to be intracellular. NHE7 (human 725 aa, chromosome Xp11.4) is ubiquitously expressed, and predominantly localizes to the trans-golgi network. NHE7 mediates the influx of Na+ or K+ in exchange for H+. It is ~70% related to NHE6 but relatively less (~25%) homologous with other NHEs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC9A7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC9A7
Gene Accession No. Q8BLV3, Q96T83
Gene Alias A530087D17Rik; hypothetical protein LOC554532; Na(+)/H(+) exchanger 7; Nhe7; NHE-7; nonselective sodium potassium/proton exchanger; SLC9A6; Slc9a7; Sodium/hydrogen exchanger 7; solute carrier family 9 (sodium/hydrogen exchanger), isoform 7; solute carrier family 9 (sodium/hydrogen exchanger), member 7; solute carrier family 9 member 7; solute carrier family 9 member A7; solute carrier family 9, subfamily A (NHE7, cation proton antiporter 7), member 7; wu:fk03b09; zgc:101088; zgc:113878
Gene Symbols SLC9A7
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC9A7(Accession Q96T83), corresponding to amino acid residues L535-A585.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 236727, 317170, 84679
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.