Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC9A8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588788
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588788 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588788 Supplier Invitrogen™ Supplier No. PA588788
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLMDIEDAKA HRRNKKDVNL SKTEKMGNTV ESEHLSELTE EEYEAHYIRR QDLKGFVWLD AKYLNPFFTR RLTQEDLHHG RIQMKTLTNK WYEEVRQGPS GSEDDEQELL; Positive Samples: 293T; Cellular Location: Golgi apparatus membrane, Multi-pass membrane protein.

Sodium-hydrogen exchangers (NHEs), such as SLC9A8, are integral transmembrane proteins that exchange extracellular Na+ for intracellular H+. NHEs have multiple functions, including intracellular pH homeostasis, cell volume regulation, and electroneutral NaCl absorption in epithelia (Xu et al., 2008).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC9A8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC9A8
Gene Accession No. Q9Y2E8
Gene Alias 1200006P13Rik; 6430709P13Rik; AI182282; DKFZp686C03237; fj61b02; FLJ42500; hypothetical protein LOC494043; KIAA0939; MGC138418; Na(+)/H(+) exchanger 8; Nhe8; NHE-8; NHE-8 antibody; SLC9A8; SLC9A8 antibody; sodium hydrogen exchanger 8; sodium/hydrogen exchanger 8; solute carrier family 9 (sodium/hydrogen exchanger), isoform 8; solute carrier family 9 (sodium/hydrogen exchanger), member 8; solute carrier family 9 member 8; solute carrier family 9 member A8; solute carrier family 9, subfamily A (NHE8, cation proton antiporter 8), member 8; wu:fj61b02; zgc:103660
Gene Symbols SLC9A8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 472-581 of human SLC9A8 (NP_0560811).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23315
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.