Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC9A9 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen SLC9A9
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1599020
SDP
View Documents
Novus Biologicals
NBP15990120UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP159901
SDP
View Documents
Novus Biologicals
NBP159901
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

SLC9A9 Polyclonal specifically detects SLC9A9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SLC9A9
Polyclonal
Rabbit
Q8IVB4
285195
Synthetic peptides corresponding to SLC9A9(solute carrier family 9 (sodium/hydrogen exchanger), member 9) The peptide sequence was selected from the C terminal of SLC9A9. Peptide sequence INYQEQASSPCSPPARLGLDQKASPQTPGKENIYEGDLGLGGYELKLEQT.
Primary
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
AUTS16, FLJ35613, Na(+)/H(+) exchanger 9, NHE9, NHE-9, putative protein product of Nbla00118, sodium/hydrogen exchanger 9, sodium/proton exchanger NHE9, solute carrier family 9 (sodium/hydrogen exchanger), solute carrier family 9 (sodium/hydrogen exchanger), isoform 9, solute carrier family 9 (sodium/hydrogen exchanger), member 9, Solute carrier family 9 member 9
SLC9A9
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.