Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC9A9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101895 Supplier Invitrogen™ Supplier No. PA5101895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC9A9.

This gene encodes a sodium hydrogen exchanger that is a member of the solute carrier 9 protein family. The encoded protein localizes the to the late recycling endosomes and may play an important role in maintaining cation homeostasis. Defects in this gene are associated with attention-deficit/hyperactivity disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC9A9
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC9A9
Gene Accession No. Q8BZ00, Q8IVB4
Gene Alias 5730527A11Rik; 9930105B05; AI854429; AUTS16; FLJ35613; Na(+)/H(+) exchanger 9; Nbla00118; NHE9; NHE-9; putative protein product of Nbla00118; RGD1560736; Slc9a9; Sodium/hydrogen exchanger 9; sodium/proton exchanger NHE9; solute carrier family 9 (sodium/hydrogen exchanger), isoform 9; solute carrier family 9 (sodium/hydrogen exchanger), member 9; solute carrier family 9 member 9; solute carrier family 9 member A9; solute carrier family 9, subfamily A (NHE9, cation proton antiporter 9), member 9
Gene Symbols SLC9A9
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC9A9(Accession Q8IVB4), corresponding to amino acid residues I172-I222.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285195, 331004
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.