Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC9A9 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985846
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5846 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5846 Supplier Bioss Supplier No. BS4031R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a sodium hydrogen exchanger that is a member of the solute carrier 9 protein family. The encoded protein localizes the to the late recycling endosomes and may play an important role in maintaining cation homeostasis. Defects in this gene are associated with attention-deficit/hyperactivity disorder.

Specifications

Antigen SLC9A9
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC9A9
Gene Accession No. Q8IVB4
Gene Alias 5730527A11Rik; 9930105B05; AI854429; AUTS16; BOS_1261; FLJ35613; LOC100355467; Na(+)/H(+) exchanger 9; Nbla00118; Nhe9; NHE-9; putative protein product of Nbla00118; RGD1560736; Slc9a9; Sodium/hydrogen exchanger 9; sodium/proton exchanger NHE9; solute carrier family 9 (sodium/hydrogen exchanger), isoform 9; solute carrier family 9 (sodium/hydrogen exchanger), member 9; Solute carrier family 9 member 9; solute carrier family 9 member A9; solute carrier family 9, member 7-like; solute carrier family 9, subfamily A (NHE9, cation proton antiporter 9), member 9
Gene Symbols SLC9A9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SLC9A9.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285195
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.