Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC9B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566225
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566225 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566225 Supplier Invitrogen™ Supplier No. PA566225
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGILGPKMLTR HYDPSKIKLQ Highest antigen sequence identity to the following orthologs - mouse 43%, rat 43%.

Sperm-specific Na(+)/H(+) exchanger involved in intracellular pH regulation of spermatozoa. Involved in sperm motility and fertility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC9B1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC9B1
Gene Accession No. Q4ZJI4
Gene Alias Na(+)/H(+) exchanger-like domain-containing protein 1; NHA1; NHE domain-containing protein 1; NHEDC1; SLC9B1; Sodium/hydrogen exchanger 9B1; sodium/hydrogen exchanger-like domain-containing protein 1; solute carrier family 9 member B1; solute carrier family 9, subfamily B (cation proton antiporter 2), member 1; solute carrier family 9, subfamily B (NHA1, cation proton antiporter 1), member 1; Solute carrier family 9, subfamily B member 1; testicular tissue protein Li 127
Gene Symbols SLC9B1
Host Species Rabbit
Immunogen Recombinant Human SLC9B1. Recombinant protein control fragment (Product #RP-106905).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150159
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.