Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC9B1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566777
Description
Immunogen sequence: VLYTMVLQENG YGVEEDIPTL LMAAS Highest antigen sequence identity to the following orthologs - mouse 65%, rat 65%.
Sperm-specific Na(+)/H(+) exchanger involved in intracellular pH regulation of spermatozoa. Involved in sperm motility and fertility.
Specifications
SLC9B1 | |
Polyclonal | |
Unconjugated | |
SLC9B1 | |
Na(+)/H(+) exchanger-like domain-containing protein 1; NHA1; NHE domain-containing protein 1; NHEDC1; SLC9B1; Sodium/hydrogen exchanger 9B1; sodium/hydrogen exchanger-like domain-containing protein 1; solute carrier family 9 member B1; solute carrier family 9, subfamily B (cation proton antiporter 2), member 1; solute carrier family 9, subfamily B (NHA1, cation proton antiporter 1), member 1; Solute carrier family 9, subfamily B member 1; testicular tissue protein Li 127 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
150159 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q4ZJI4 | |
SLC9B1 | |
Recombinant Human SLC9B1. Recombinant protein control fragment (Product #RP-107435). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction