Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC9B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569807
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569807 Supplier Invitrogen™ Supplier No. PA569807
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

Sperm-specific Na(+)/H(+) exchanger involved in intracellular pH regulation of spermatozoa. Involved in sperm motility and fertility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC9B1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC9B1
Gene Accession No. Q4ZJI4
Gene Alias Na(+)/H(+) exchanger-like domain-containing protein 1; NHA1; NHE domain-containing protein 1; NHEDC1; SLC9B1; Sodium/hydrogen exchanger 9B1; sodium/hydrogen exchanger-like domain-containing protein 1; solute carrier family 9 member B1; solute carrier family 9, subfamily B (cation proton antiporter 2), member 1; solute carrier family 9, subfamily B (NHA1, cation proton antiporter 1), member 1; Solute carrier family 9, subfamily B member 1; testicular tissue protein Li 127
Gene Symbols SLC9B1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human SLC9B1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150159
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.