Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLCO1B7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569325
Description
This target displays homology in the following species: Human: 100%; Rat: 91%.
SLCO1B7 gene ontology annotations related to this gene include bile acid and bile salt transport; sodium-independent organic anion transport.
Specifications
SLCO1B7 | |
Polyclonal | |
Unconjugated | |
SLCO1B7 | |
liver-specific organic anion transporter 3; liver-specific organic anion transporter 3TM12; LST3; LST-3; LST3TM12; LST-3TM12; organic anion transporter LST-3b; putative solute carrier organic anion transporter family member 1B7; SLC21A21; SLCO1B7; solute carrier organic anion transporter family member 1B7 (non-functional); solute carrier organic anion transporter family member 1B7 (putative); solute carrier organic anion transporter family, member 1B7 (non-functional) | |
Rabbit | |
Affinity chromatography | |
RUO | |
338821 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
G3V0H7 | |
SLCO1B7 | |
synthetic peptide directed towards the middle region of human SLCO1B7. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction