Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLCO1C1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115919 Supplier Invitrogen™ Supplier No. PA5115919
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLCO1C1.

This gene encodes a member of the organic anion transporter family. The encoded protein is a transmembrane receptor that mediates the sodium-independent uptake of thyroid hormones in brain tissues. This protein has particularly high affinity for the thyroid hormones thyroxine, tri-iodothyronine and reverse tri-iodothyronine. Polymorphisms in the gene encoding this protein may be associated with fatigue and depression in patients suffering from hyperthyroidism. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLCO1C1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene SLCO1C1
Gene Accession No. Q9NYB5
Gene Alias BBB-specific anion transporter type 1; blood-brain barrier specific anion transporter; blood-brain barrier-specific anion transporter 1; Bsat1; OATP1; Oatp14; OATP-14; OATP1C1; Oatp2; OATPF; OATP-F; OATPRP5; OAT-RP-5; Organic anion transporter 2; organic anion transporter F; Organic anion transporter polypeptide-related protein 5; organic anion transporting polypeptide 14; organic anion-transporting polypeptide 14; Slc21a14; SLCO1C1; solute carrier family 21 (organic anion transporter), member 14; solute carrier family 21 member 14; Solute carrier organic anion transporter family member 1C1; solute carrier organic anion transporter family, member 1C1; thyroxine transporter
Gene Symbols SLCO1C1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLCO1C1(Accession Q9NYB5), corresponding to amino acid residues R673-L712.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53919
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.