Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLCO2B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554258
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554258 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554258 Supplier Invitrogen™ Supplier No. PA554258
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GITHQTSAHP GLELSPSCME ACSCPLDGFN PVCDPSTRVE YITPCHAGCS SWVVQDALDN SQVFYTNCSC VVEGNPVLAG SCDSTCSHLV VPF Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.

SLCO2B1 mediates the Na+-independent transport of organic anions such as taurocholate, the prostaglandins PGD2, PGE1, PGE2, leukotriene C4, thromboxane B2 and iloprost.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLCO2B1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLCO2B1
Gene Accession No. O94956
Gene Alias AI060904; AI852653; hypothetical protein LOC557480; im:7158298; KIAA0880; moat1; multispecific organic anion transporter 1; Oatp2b1; OATPB; OATP-B; OATP-RP2; Organic anion transporter B; organic anion transporter moatp1; Organic anion transporter polypeptide-related protein 2; organic anion transporting polypeptide 2b1; organic anion-transporting polypeptide 2B1; SLC21A9; SLCO/OATP2B1; Slco2b1; solute carrier anion transporting family member 2b1; solute carrier family 21 (organic anion transporter), member 9; solute carrier family 21 member 9; Solute carrier organic anion transporter family member 2B1; solute carrier organic anion transporter family, member 2B1; wu:fi40d02; zgc:123236
Gene Symbols SLCO2B1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLCO2B1. Recombinant protein control fragment (Product #RP-101764).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11309
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.