Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLCO2B1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587616
Description
Immunogen sequence: LLMTLPHFIS EPYRYDNTSP EDMPQDFKAS LCLPTTSAPA SAPSNGNCSS YTETQHLSVV GIMFVAQTLL GVGGVPIQPF G; Positive Samples: HepG2, Mouse liver; Cellular Location: Cell membrane, Multi-pass membrane protein.
SLCO2B1 mediates the Na+-independent transport of organic anions such as taurocholate, the prostaglandins PGD2, PGE1, PGE2, leukotriene C4, thromboxane B2 and iloprost.
Specifications
SLCO2B1 | |
Polyclonal | |
Unconjugated | |
SLCO2B1 | |
AI060904; AI852653; hypothetical protein LOC557480; im:7158298; KIAA0880; moat1; multispecific organic anion transporter 1; Oatp2b1; OATPB; OATP-B; OATP-RP2; Organic anion transporter B; organic anion transporter moatp1; Organic anion transporter polypeptide-related protein 2; organic anion transporting polypeptide 2b1; organic anion-transporting polypeptide 2B1; SLC21A9; SLCO/OATP2B1; Slco2b1; solute carrier anion transporting family member 2b1; solute carrier family 21 (organic anion transporter), member 9; solute carrier family 21 member 9; Solute carrier organic anion transporter family member 2B1; solute carrier organic anion transporter family, member 2B1; wu:fi40d02; zgc:123236 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
101488, 11309 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O94956, Q8BXB6 | |
SLCO2B1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 130-210 of human SLCO2B1 (NP_0091871). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction