Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLCO2B1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542453
Description
Peptide sequence: DPQDVRPSVF HNIKLFVLCH SLLQLAQLMI SGYLKSSIST VEKRFGLSSQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
SLCO2B1 mediates the Na+-independent transport of organic anions such as taurocholate, the prostaglandins PGD2, PGE1, PGE2, leukotriene C4, thromboxane B2 and iloprost.
Specifications
SLCO2B1 | |
Polyclonal | |
Unconjugated | |
SLCO2B1 | |
AI060904; AI852653; hypothetical protein LOC557480; im:7158298; KIAA0880; moat1; multispecific organic anion transporter 1; Oatp2b1; OATPB; OATP-B; OATP-RP2; Organic anion transporter B; organic anion transporter moatp1; Organic anion transporter polypeptide-related protein 2; organic anion transporting polypeptide 2b1; organic anion-transporting polypeptide 2B1; SLC21A9; SLCO/OATP2B1; Slco2b1; solute carrier anion transporting family member 2b1; solute carrier family 21 (organic anion transporter), member 9; solute carrier family 21 member 9; Solute carrier organic anion transporter family member 2B1; solute carrier organic anion transporter family, member 2B1; wu:fi40d02; zgc:123236 | |
Rabbit | |
Affinity chromatography | |
RUO | |
11309 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O94956 | |
SLCO2B1 | |
Synthetic peptide directed towards the N-terminal of human SLCO2B1 (aa 36-85). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction