Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLCO3A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542457
Description
Peptide sequence: MEIAVVAGFA AFLGKYLEQQ FNLTTSSANQ LLGMTAIPCA CLGIFLGGLL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
SLCO3A1 mediates the Na+-independent transport of organic anions such as estrone-3-sulfate. It mediates transport of prostaglandins (PG) E1 and E2, thyroxine (T4), deltorphin II, BQ-123 and vasopressin, but not DPDPE (a derivative of enkephalin lacking an N-terminal tyrosine residue), estrone-3-sulfate, taurocholate, digoxin nor DHEAS.
Specifications
SLCO3A1 | |
Polyclonal | |
Unconjugated | |
SLCO3A1 | |
5830414C08Rik; androgen regulated protein 1; Anr1; FLJ40478; LOW QUALITY PROTEIN: solute carrier organic anion transporter family member 3A1; MJAM; OATP3A1; OATPD; OATP-D; OATPRP3; OATP-RP3; Organic anion transporter polypeptide-related protein 3; organic anion transporting polypeptide 3a1; organic anion-transporting polypeptide D; PGE1 transporter; Pgt2; Prostaglandin transporter subtype 2; R75096; si:ch211-196l7.1; SLC21A11; Slco3a1; Sodium-independent organic anion transporter D; solute carrier family 21 (organic anion transporter), member 11; solute carrier family 21 member 11; Solute carrier organic anion transporter family member 3A1; solute carrier organic anion transporter family, member 3A1; wu:fa93e11; zOatp3a2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
28232 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UIG8 | |
SLCO3A1 | |
Synthetic peptide directed towards the middle region of human SLCO3A1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction