Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLCO4A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556536
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556536 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556536 Supplier Invitrogen™ Supplier No. PA556536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GYPRQLPGSQ RYAVMRAAEM HQLKDSSRGE ASNPDFGKTI RDLPL Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 78%.

Mediates the Na(+)-independent transport of organic anions such as the thyroid hormones T3 (triiodo-L-thyronine), T4 (thyroxine) and rT3, and of estrone-3-sulfate and taurocholate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLCO4A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLCO4A1
Gene Accession No. Q96BD0
Gene Alias colon organic anion transporter; OATP1; OATP4A1; OATPE; OATP-E; OATPRP1; OATP-RP1; Organic anion transporter polypeptide-related protein 1; organic anion transporting polypeptide E; organic anion-transporting polypeptide E; POAT; SLC21A12; SLCO4A1; Sodium-independent organic anion transporter E; solute carrier family 21 (organic anion transporter), member 12; solute carrier family 21 member 12; Solute carrier organic anion transporter family member 4A1; solute carrier organic anion transporter family, member 4A1
Gene Symbols SLCO4A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLCO4A1. Recombinant protein control fragment (Product #RP-95579).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28231
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.