Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLCO4C1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306278AF750
Description
SLCO4C1 Polyclonal specifically detects SLCO4C1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SLCO4C1 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
PRO2176, solute carrier family 21 member 20, solute carrier organic anion transporter family, member 4C1 | |
Produced in rabbits immunized with E. coli-derived Human SLCO4C1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
353189 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction