Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLCO4C1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112274
Description
SLCO4C1 Polyclonal Antibody for ICC/IF, IHC (P)
Organic anion transporter, capable of transporting pharmacological substances such as digoxin, ouabain, thyroxine, methotrexate and cAMP. May participate in the regulation of membrane transport of ouabain. Involved in the uptake of the dipeptidyl peptidase-4 inhibitor sitagliptin and hence may play a role in its transport into and out of renal proximal tubule cells. May be involved in the first step of the transport pathway of digoxin and various compounds into the urine in the kidney. May be involved in sperm maturation by enabling directed movement of organic anions and compounds within or between cells. This ion- transporting process is important to maintain the strict epididymal homeostasis necessary for sperm maturation. May have a role in secretory functions since seminal vesicle epithelial cells are assumed to secrete proteins involved in decapacitation by modifying surface proteins to facilitate the acquisition of the ability to fertilize the egg.
Specifications
SLCO4C1 | |
Polyclonal | |
Unconjugated | |
SLCO4C1 | |
9530051F04; C330017E21Rik; Oatp4c1; OATP-H; OATP-M1; oatp-R; OATPX; Organic anion transporter M1; PRO2176; SLC21A20; SLCO4C1; solute carrier family 21 member 20; Solute carrier organic anion transporter family member 4C1; solute carrier organic anion transporter family, member 4C1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
353189 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 0.03% ProClin 300; pH 7 | |
Q6ZQN7 | |
SLCO4C1 | |
E. coli-derived Human SLCO4C1 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction