Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLCO4C1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140093
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140093 Supplier Invitrogen™ Supplier No. PA5140093
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PKHLPGTAEI QAGKTSQAHQ SNSNADVKFG KSIKDFPAAL KNLMK Highest antigen sequence indentity to the following orthologs: Rat - 71%, Mouse - 71%.

Organic anion transporter, capable of transporting pharmacological substances such as digoxin, ouabain, thyroxine, methotrexate and cAMP. May participate in the regulation of membrane transport of ouabain. Involved in the uptake of the dipeptidyl peptidase-4 inhibitor sitagliptin and hence may play a role in its transport into and out of renal proximal tubule cells. May be involved in the first step of the transport pathway of digoxin and various compounds into the urine in the kidney. May be involved in sperm maturation by enabling directed movement of organic anions and compounds within or between cells. This ion- transporting process is important to maintain the strict epididymal homeostasis necessary for sperm maturation. May have a role in secretory functions since seminal vesicle epithelial cells are assumed to secrete proteins involved in decapacitation by modifying surface proteins to facilitate the acquisition of the ability to fertilize the egg.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLCO4C1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLCO4C1
Gene Accession No. Q6ZQN7
Gene Alias 9530051F04; C330017E21Rik; Oatp4c1; OATP-H; OATP-M1; oatp-R; OATPX; Organic anion transporter M1; PRO2176; SLC21A20; SLCO4C1; solute carrier family 21 member 20; Solute carrier organic anion transporter family member 4C1; solute carrier organic anion transporter family, member 4C1
Gene Symbols SLCO4C1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLCO4C1. Recombinant protein control fragment (Product #RP-108958).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353189
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.