Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLCO5A1 Polyclonal Antibody
SLCO5A1 Polyclonal Antibody
Supplier: Thermo Scientific PA555218
Description
SLCO5A1 Polyclonal Antibody for IHC

Specifications
SLCO5A1 | |
Polyclonal | |
Unconjugated | |
SLCO5A1 | |
4732454B15; A630033C23Rik; AA407151; LOW QUALITY PROTEIN: solute carrier organic anion transporter family member 5A1; OATP5A1; OATPJ; OATP-J; OATPRP4; OATP-RP4; organic anion transporter polypeptide-related protein 4; SLC21A15; SLCO5A1; solute carrier family 21 (organic anion transporter), member 15; solute carrier family 21 member 15; Solute carrier organic anion transporter family member 5A1; solute carrier organic anion transporter family, member 5A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
81796 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H2Y9 | |
SLCO5A1 | |
Recombinant protein corresponding to Human SLCO5A1. Recombinant protein control fragment (Product #RP-93798). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction