Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLCO6A1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | SLCO6A1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
SLCO6A1 Polyclonal specifically detects SLCO6A1 in Human samples. It is validated for Western Blot.Specifications
SLCO6A1 | |
Polyclonal | |
Purified | |
RUO | |
Cancer/testis antigen 48, CT48Organic anion-transporting polypeptide I, Gonad-specific transporter, GSTOATP-I, MGC26949, OATP6A1Solute carrier family 21 member 19, OATPY, Organic anion-transporting polypeptide 6A1, SLC21A19, solute carrier organic anion transporter family member 6A1, solute carrier organic anion transporter family, member 6A1, testis-specific organic anion transporter | |
SLCO6A1 | |
IgG | |
Protein A purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Q86UG4 | |
133482 | |
Synthetic peptides corresponding to SLCO6A1(solute carrier organic anion transporter family, member 6A1) The peptide sequence was selected from the C terminal of SLCO6A1. Peptide sequence LAMTRVVPDKLRSLALGVSYVILRIFGTIPGPSIFKMSGETSCILRDVNK. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title