Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLCO6A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542520
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542520 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542520 Supplier Invitrogen™ Supplier No. PA542520
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CCNNIRCFMI FYCILLICQG VVFGLIDVSI GDFQKEYQLK TIEKLALEKS Sequence homology: Human: 100%.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLCO6A1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLCO6A1
Gene Accession No. Q86UG4
Gene Alias cancer/testis antigen 48; CT48; Gonad-specific transporter; GST; OATP6A1; OATP-I; OATPY; Organic anion-transporting polypeptide 6A1; Organic anion-transporting polypeptide I; SLC21A19; SLCO6A1; Solute carrier family 21 member 19; solute carrier organic anion transporter family member 6A1; solute carrier organic anion transporter family, member 6A1; testicular tissue protein Li 174; testis-specific organic anion transporter
Gene Symbols SLCO6A1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLCO6A1 (aa 101-150).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133482
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.