Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLCO6A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562926 Supplier Invitrogen™ Supplier No. PA562926
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGCTYSKAQN QKKMYYNCSC IKEGLITADA EGDFIDARPG KCDAKCYKLP L Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 63%.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLCO6A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLCO6A1
Gene Accession No. Q86UG4
Gene Alias cancer/testis antigen 48; CT48; Gonad-specific transporter; GST; OATP6A1; OATP-I; OATPY; Organic anion-transporting polypeptide 6A1; Organic anion-transporting polypeptide I; SLC21A19; SLCO6A1; Solute carrier family 21 member 19; solute carrier organic anion transporter family member 6A1; solute carrier organic anion transporter family, member 6A1; testicular tissue protein Li 174; testis-specific organic anion transporter
Gene Symbols SLCO6A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLCO6A1. Recombinant protein control fragment (Product #RP-101613).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133482
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.