Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sleeping Beauty Transposase Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Supplier: R&D Systems AF2798
This item is not returnable.
View return policy
Description
Sleeping Beauty Transposase Polyclonal specifically detects Sleeping Beauty Transposase in Multi-Species samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Sleeping Beauty Transposase | |
Polyclonal | |
Unconjugated | |
Goat | |
Affinity Purified | |
RUO | |
Detects Sleeping Beauty Transposase in direct ELISAs and Western blots. | |
Multi-species | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
E. coli-derived recombinant Sleeping Beauty Transposase | |
100 μg | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction