Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLFN11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566453
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566453 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566453 Supplier Invitrogen™ Supplier No. PA566453
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ETSVRSMDSRE AFCFLKTKRK PKILEEGPFH KIHKGVYQEL PNSDPADPNS DPADLIFQKD YLEYGEILP Highest antigen sequence identity to the following orthologs - mouse 41%, rat 39%.

Inhibitor of DNA replication that promotes cell death in response to DNA damage (PubMed:22927417, PubMed:26658330, PubMed:29395061). Acts as a guardian of the genome by killing cells with defective replication (PubMed:29395061). Persistently blocks stressed replication forks by opening chromatin across replication initiation sites at stressed replication forks, possibly leading to unwind DNA ahead of the MCM helicase and block fork progression, ultimately leading to cell death (PubMed:29395061). Acts independently of ATR (PubMed:29395061). Also acts as an interferon (IFN)-induced antiviral protein which acts as an inhibitor of retrovirus protein synthesis (PubMed:23000900). Specifically abrogates the production of retroviruses such as human immunodeficiency virus 1 (HIV-1) by acting as a specific inhibitor of the synthesis of retroviruses encoded proteins in a codon-usage-dependent manner (PubMed:23000900). Binds to tRNAs and exploits the unique viral codon bias towards A/T nucleotides (PubMed:23000900). The exact inhibition mechanism is unclear: may either sequester tRNAs, prevent their maturation via post-transcriptional processing or may accelerate their deacylation (PubMed:23000900). Does not inhibit reverse transcription, integration or production and nuclear export of viral RNA (PubMed:23000900). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLFN11
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLFN11
Gene Accession No. Q7Z7L1
Gene Alias schlafen family member 11; SLFN11; SLFN8/9
Gene Symbols SLFN11
Host Species Rabbit
Immunogen Recombinant Human SLFN11. Recombinant protein control fragment (Product #RP-107126).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91607
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.