Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLFN12L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555080
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555080 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555080 Supplier Invitrogen™ Supplier No. PA555080
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NRVKQLTEKE WIQFMVDSEP VCEELPSPAS TSSPVSQSYP LREYINFKIQ PLRYHLPGLS EKITCAPKTF CRNLFSQHEG LK Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 30%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SLFN12L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLFN12L
Gene Accession No. Q6IEE8
Gene Alias schlafen family member 12 like; schlafen family member 12-like; SLFN12L; SLFN5
Gene Symbols SLFN12L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLFN12L. Recombinant protein control fragment (Product #RP-93562).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100506736
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.