Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLFN14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554658
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554658 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554658 Supplier Invitrogen™ Supplier No. PA554658
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VFSLPLRICS LRSNLYRRDV TSAINLSASS ALELLREKGF RAQRGRPRVK KLHPQQVLNR CIQEEEDMRI LASEFFKKDK LMYKEKLNF Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 73%.

Despite being first described several years ago, the roles of the Schlafen (SLFN) family of proteins remain largely unknown. The SLFN genes are preferentially expressed in lymphoid tissues and differentially regulated during thymocyte maturation. It is thought that many play roles in cell growth, hemopoietic cell differentiation, and T cell development and maturation. Most members contain at least one divergent AAA domain (AAA_4) that may play a role in ATP binding. SLFN14 is a recently identified member of the SLFN family; its role has yet to be determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLFN14
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLFN14
Gene Accession No. P0C7P3
Gene Alias BDPLT20; C-terminally truncated SLFN14 endoribonuclease; Gm20; LRRGT00163; Protein SLFN14; schlafen 14; schlafen family member 14; schlafen family member 14, pseudogene; SLFN14; Slfn14-ps
Gene Symbols SLFN14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLFN14. Recombinant protein control fragment (Product #RP-93547).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342618
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.