Learn More
Invitrogen™ SLFN14 Polyclonal Antibody

Description
A suggested positive control is mouse kidney tissue lysate. PA5-20868 can be used with blocking peptide PEP-0982.
Specifications
Specifications
| Antigen | SLFN14 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SLFN14 |
| Gene Accession No. | P0C7P3, V9GXG1 |
| Gene Alias | BDPLT20; C-terminally truncated SLFN14 endoribonuclease; Gm20; LRRGT00163; Protein SLFN14; schlafen 14; schlafen family member 14; schlafen family member 14, pseudogene; SLFN14; Slfn14-ps |
| Gene Symbols | SLFN14 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.