Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Slfn9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144357
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144357 Supplier Invitrogen™ Supplier No. PA5144357
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Slfn9 Polyclonal Antibody for Western Blot, ELISA

Endoribonuclease that cleaves tRNAs and rRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Slfn9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Slfn9
Gene Accession No. B1ARD6
Gene Alias EC 3.1.-.-; Schlafen family member 9; Schlafen-9; Slfn9
Gene Symbols Slfn9
Host Species Rabbit
Immunogen Recombinant Mouse Schlafen family member 9 protein (1-83AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237886
Target Species Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.