Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLFNL1 Antibody (OTI1G2), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274223G
Description
SLFNL1 Monoclonal specifically detects SLFNL1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SLFNL1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SLFNL1 | |
Full length recombinant protein of human SLFNL1 (NP_659427) produced in HEK293T cells. | |
0.1 mL | |
200172.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1G2 | |
Western Blot, Immunohistochemistry | |
FLJ23878, MGC43873, schlafen-like 1, schlafen-like protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction