Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLFNL1 Monoclonal Antibody (OTI5C3), TrueMAB™, OriGene
SDP

Catalog No. 502541857
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1857 30 μL
50-167-3104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1857 Supplier OriGene Supplier No. TA502017S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLFNL1 Monoclonal antibody specifically detects SLFNL1 in Human samples. It is validated for Western Blot

Specifications

Antigen SLFNL1
Applications Western Blot
Classification Monoclonal
Clone OTI5C3
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SDR9C7
Gene Accession No. Q8NEX9
Gene Alias 4933406A14Rik; schlafen like 1; schlafen-like 1; schlafen-like protein 1; SLFNL1; testicular tissue protein Li 173
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-313 of human SDR9C7 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121214
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.