Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLFNL1 Mouse anti-Human, Clone: OTI5C3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502017

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SLFNL1 belongs to the Schlafen family. The function of the SLFNL1 protein remains unknown. There are four different isoforms.Spécifications
SLFNL1 | |
Monoclonal | |
Unconjugated | |
SLFNL1 | |
schlafen like 1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
200172 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5C3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q499Z3 | |
SLFNL1 | |
Full length human recombit protein of human SLFNL1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
missing translation for 'provideContentCorrection'
missing translation for 'type07FormHeader'
missing translation for 'productTitle'
missing translation for 'spotOppurtunityImprovement'missing translation for 'shareContentCorrection'