Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 10K MWCO

Catalog No. pi66380
Click to view available options
Sample Volume (Metric):
0.5 mL
3 mL
12 mL
30 mL
Quantity:
10 Cassettes
5 x 10 Cassettes
5 x 8 Cassettes
6 Cassettes
8 Cassettes
Product Type:
Dialysis Cassette Kit
Dialysis Cassettes
Dialysis Cassettes (gamma-irradiated)

Remove salts and small contaminants to maximize sample purity and recovery with Slide-A-Lyzer 10K MWCO dialysis cassettes.

Slide-A-Lyzer Dialysis Cassettes (10K MWCO) facilitate the simple and effective removal of salts and small contaminants from proteins and other macromolecules greater than 10,000 Daltons. Slide-A-Lyzer Dialysis Cassettes are available in four sizes to accommodate sample volumes between 0.5 and 30 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange and more complete sample recovery.

Slide-A-Lyzer cassettes can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.

Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

Features of Slide-A-Lyzer Dialysis Cassettes:
Color-coded—orange frame specifies cassettes with the 10,000 MW pore size membrane
Four cassette sizes—select the cassette that best suits your sample volume
Fast dialysis—flat cassette chamber with two membranes provides high surface area-to-volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
High recovery—rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports
Gamma-irradiated option—gamma-irradiated cassettes are available for applications requiring conditions that minimize the risk of sample contamination

Gamma-irradiated dialysis cassettes
Gamma-irradiated Slide-A-Lyzer Dialysis Cassettes are ideal for researchers culturing cells and microorganisms; purifying viruses, DNA, and RNA; or performing sample preparation for other applications requiring conditions that minimize the risk of sample contamination. The gamma-irradiated cassettes are treated with 20–36 kGy of gamma radiation, a dosage that has been determined to sterilize the units based on both trypticase soy broth medium for identifying total growth and fluid thioglycollate medium for identifying anaerobic growth. However, a confirmation of sterility is not provided for individual lots, as no further sterility testing is conducted post-irradiation.

Membrane specifications:
Composition: regenerated cellulose synthesized by the viscose method
Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
Hydration time required before use: 2 minutes
Glycerol content: 21%
Sulfur content: 0.1–0.15%
Heavy metals content: trace

Volume ranges:
• 0.5 mL = 0.1 to 0.5 mL
• 3 mL = 0.5 to 3 mL
• 12 mL = 3 to 12 mL
• 30 mL = 12 to 30 mL

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
MWCO 10 kDa
Product Line Slide-A-Lyzer™
Quantity 10 Cassettes
Sample Volume (Metric) 3 mL
Product Type Dialysis Cassettes
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.