Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Cassette Float Buoys (for 12 mL cassettes), 8 Buoys PROMO

Catalog No. PI66432PM
Click to view available options
:
Thermo Scientific™ Slide-A-Lyzer™ Cassette Float Buoys

Slide-A-Lyzer Float Buoys and Syringes are available to support a variety of dialysis cassettes for volumes of 0.1 to 20 mL.

Minimize sample loss into and out of Slide-A-Lyzer Dialysis Cassettes using Slide-A-Lyzer Dialysis Cassette Syringes for 1-mL to 20-mL sample volumes. Slide-A-Lyzer Cassette Float Buoys are designed for floating 0.5- to 12-mL cassettes in dialysate.

Features of Slide-A-Lyzer Cassette Accessories include:

  • Syringes are appropriately sized for different sample volumes
  • Buoys keep stir bar from tearing dialysis membrane
  • Matched to Slide-A-Lyzer Cassette for a secure fit and minimization of sample loss
  • Chemically inert

Specifications

Sufficient For Floating one 12 mL Cassette with each buoy
Content And Storage Store at room temperature.
Membrane Dialysis
Format Cassette, Accessory
Capacity (Metric) For 12 mL Cassettes
Product Line Slide-A-Lyzer™
Quantity 8 Buoys
Sample Volume (Metric) 12 mL
Product Type Cassette Float Buoy
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.