Learn More
Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 2K MWCO, 3 mL, 10 Cassettes PROMO
Description
Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. Slide-A-Lyzer cassettes can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers. No knots, clips, or caps are needed to seal the units and prevent leaking and sample loss during the dialysis procedure.
Features of Slide-A-Lyzer Dialysis Cassettes:
- Color-coded—blue frame specifies cassettes with the 2,000-MW pore-size membrane
- Four cassette sizes—select the cassette that best suits your sample volume (0.5 to 30 mL)
- Fast dialysis—flat cassette chamber with two membranes provides high surface area-to-volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
- High recovery—rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports
Membrane specifications:
- Composition: regenerated cellulose synthesized by the viscose method
- Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
- Hydration time required before use: 2 minutes for normal volumes or 4 minutes for minimal volumes
- Glycerol content: none
- Sulfur content: 0.169%
- Heavy metals content: trace
Volume ranges:
- 0.5 mL = 0.1 to 0.5 mL
- 3 mL = 0.5 to 3 mL
- 12 mL = 3 to 12 mL
- 30 mL = 12 to 30 mL
Specifications
Specifications
Content And Storage | Store at room temperature |
Purification Target | Buffer Exchange, Protein |
Membrane | Cellulose Membrane, Dialysis |
Format | Cassette |
MWCO | 2.0 kDa |
Product Line | Slide-A-Lyzer™ |
Quantity | 10 Cassettes |
Sample Volume (Metric) | 3 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.