Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 3.5K MWCO, 12 mL, 8 Cassettes PROMO

Catalog No. PI66110PM
Click to view available options
:
Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassette Kit

Remove salts and contaminants, and maximize sample recovery using this semi-permeable membrane with a 3,500-Dalton protein MW cut-off (MWCO) cassette that is easier to use than traditional dialysis tubing.

Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample loss during the dialysis procedure. Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

Features of Slide-A-Lyzer Dialysis Cassettes:

  • Easy to use—no knots or clamps are needed, just inject sample into cassette and begin dialysis
  • Fast dialysis—flat cassette chamber with two membranes provides high surface area-to-volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
  • High recovery—rectangular cassette design maximizes recovery of entire sample volume from any one of the four corner injection ports
  • Four cassette sizes—select the cassette that best suits your sample volume (0.5 to 30 mL)
  • Color-coded—pink cassette frame specifies cassettes with the 3,500 MW pore size membrane

Membrane specifications:

  • Composition: regenerated cellulose synthesized by the viscose method
  • Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
  • Hydration time required before use: 2 minutes
  • Glycerol content: none
  • Sulfur content: 0.1–0.15%
  • Heavy metals content: trace

Volume ranges:

  • 0.5 mL = 0.1 to 0.5 mL
  • 3 mL = 0.5 to 3 mL
  • 12 mL = 3 to 12 mL
  • 30 mL = 12 to 30 mL

Specifications

Content And Storage Store at room temperature
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
MWCO 3.5 kDa
Product Line Slide-A-Lyzer™
Quantity 8 Cassettes
Sample Volume (Metric) 12 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.