Learn More
Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 7K MWCO, 12 mL, 8 Cassettes PROMO
Description
Slide-A-Lyzer cassettes can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.
Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.
Features of Slide-A-Lyzer Dialysis Cassettes:
- Easy to use—no knots or clamps are needed; just inject sample into cassette and begin dialysis
- Fast dialysis—flat cassette chamber with two membranes provides high surface-area to volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
- High recovery—rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports
- Four cassette sizes—select the cassette that best suits your sample volume
- Color-coded—green cassette frame specifies cassettes with the 7,000 MW pore size membrane
Membrane specifications:
- Composition: regenerated cellulose synthesized by the viscose method
- Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
- Hydration time required before use: 2 minutes
- Glycerol content: 13%
- Sulfur content: 0.1 to 0.15%
- Heavy metals content: trace
Volume ranges:
- 0.5 mL = 0.1 to 0.5 mL
- 3 mL = 0.5 to 3 mL
- 12 mL = 3 to 12 mL
Specifications
Specifications
| Content And Storage | Store at room temperature |
| Purification Target | Buffer Exchange, Protein |
| Membrane | Cellulose Membrane, Dialysis |
| Format | Cassette |
| MWCO | 7.0 kDa |
| Product Line | Slide-A-Lyzer™ |
| Quantity | 8 Cassettes |
| Sample Volume (Metric) | 12 mL |
Slide-A-Lyzer Dialysis Cassette Technology is protected by U.S. Patent 5,503,741.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.