Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Flasks, 10K MWCO, 250 mL, 4 Flasks PROMO

Catalog No. PI87762PM
Click to view available options
:
Thermo Scientific™ Slide-A-Lyzer™ Dialysis Flasks

Catalog No. PI87762PM

missing translation for 'mfr': Thermo Scientific™ 87762

Only null left
Add to cart
Add to cart

Facilitate removal of salts and small contaminants from proteins or macromolecules in sample volumes up to 250 mL. Each Slide-A-Lyzer Dialysis Flask provides flotation disks that when attached to the neck of the flask, allows the flask to float in a vertical position for simple and safe handling.

The dialysis flasks are available in distinct colors, corresponding to the pore size (MWCO) of the dialysis membrane: purple (20,000 Daltons), orange (10,000 Daltons), pink (3,500 Daltons), and blue (2,000 Daltons).

Slide-A-Lyzer Dialysis Flasks are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of an inert plastic body. The membrane and flask materials are compatible with most laboratory buffers.

Features of Slide-A-Lyzer Dialysis Flasks:

  • Easy to use—simply pipette or pour sample into flask and begin dialysis
  • Fast dialysis—flat flask chamber with two membranes provides high surface area to volume ratio, enabling dialysis of a 250 mL sample in 8 hours to overnight
  • High recovery—rectangular flask design maximizes recovery of entire sample volume via opening at top of flask
  • Multiple molecular-weight cut offs—select the membrane MWCO that best suits your sample's molecular weight

Dialysis membrane specifications:

  • Composition: regenerated-cellulose
  • Thickness: 0.9–1.2 mil (22.5 to 30 μm); 2K MWCO = 2 mil (50 μm)
  • Hydration time required before use: 2 minutes
  • Glycerol content: trace (10K MWCO content is 21%)
  • Sulfur content: 0.04–0.17%
  • Heavy metals content: trace

With Slide-A-Lyzer Dialysis Flasks, typical low-molecular weight contaminant removal, buffer exchange, and desalting can be accomplished in eight hours to overnight. The rate of dialysis depends on a variety of factors, including sample volume, size and shape of the molecule being dialyzed, agitation, and temperature. Minimum dialysis time must be determined empirically for each specific system.

Specifications

Content And Storage Shipped at ambient temperature. Upon receipt, store at room temperature.
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Flask
MWCO 10 kDa
Product Line Slide-A-Lyzer™
Quantity 4 Flasks
Sample Volume (Metric) 250 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.