Learn More
Thermo Scientific™ Slide-A-Lyzer™ Dialysis Flasks, 20K MWCO, 250 mL, 4 Flasks PROMO
Description
The dialysis flasks are available in distinct colors, corresponding to the pore size (MWCO) of the dialysis membrane: purple (20,000 Daltons), orange (10,000 Daltons), pink (3,500 Daltons), and blue (2,000 Daltons).
Slide-A-Lyzer Dialysis Flasks are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of an inert plastic body. The membrane and flask materials are compatible with most laboratory buffers.
Features of Slide-A-Lyzer Dialysis Flasks:
- Easy to use—simply pipette or pour sample into flask and begin dialysis
- Fast dialysis—flat flask chamber with two membranes provides high surface area to volume ratio, enabling dialysis of a 250 mL sample in 8 hours to overnight
- High recovery—rectangular flask design maximizes recovery of entire sample volume via opening at top of flask
- Multiple molecular-weight cut offs—select the membrane MWCO that best suits your sample's molecular weight
Dialysis membrane specifications:
- Composition: regenerated-cellulose
- Thickness: 0.9–1.2 mil (22.5 to 30 μm); 2K MWCO = 2 mil (50 μm)
- Hydration time required before use: 2 minutes
- Glycerol content: trace (10K MWCO content is 21%)
- Sulfur content: 0.04–0.17%
- Heavy metals content: trace
With Slide-A-Lyzer Dialysis Flasks, typical low-molecular weight contaminant removal, buffer exchange, and desalting can be accomplished in eight hours to overnight. The rate of dialysis depends on a variety of factors, including sample volume, size and shape of the molecule being dialyzed, agitation, and temperature. Minimum dialysis time must be determined empirically for each specific system.
Specifications
Specifications
| Content And Storage | Shipped at ambient temperature. Upon receipt, store at room temperature. |
| Purification Target | Buffer Exchange, Protein |
| Membrane | Cellulose Membrane, Dialysis |
| Format | Flask |
| MWCO | 20 kDa |
| Product Line | Slide-A-Lyzer |
| Quantity | 4 Flasks |
| Sample Volume (Metric) | 250 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.