Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Flasks

Catalog No. pi87760
Change view
Click to view available options
Description:
Dialysis Flasks, 10K MWCO
Dialysis Flasks, 20K MWCO
Dialysis Flasks, 2K MWCO
Dialysis Flasks, 3.5K MWCO
Dialysis Flotation Disks
Catalog No. Description
PI87760 Dialysis Flasks, 2K MWCO
PI87759 Dialysis Flotation Disks
PI87761 Dialysis Flasks, 3.5K MWCO
PI87762 Dialysis Flasks, 10K MWCO
PI87763 Dialysis Flasks, 20K MWCO
5 options

Catalog No. PI87760

Supplier: Thermo Scientific™ 87760

Only null left
Add to Cart
Add to Cart

Facilitate removal of salts and small contaminants from proteins or macromolecules in sample volumes up to 250 mL. Each Slide-A-Lyzer Dialysis Flask provides flotation disks that when attached to the neck of the flask, allows the flask to float in a vertical position for simple and safe handling.

Slide-A-Lyzer Dialysis Flasks (2K, 3.5K, 10K, or 20K MWCO) facilitate simple and effective removal of buffer salts and small contaminants in sample volumes up to 250 mL. They are easy to use: add and remove sample using a serological pipette or by directly pouring the sample through a wide screw-cap opening at the top of the flask. No knots or clips are needed to seal the units. Slide-A-Lyzer Dialysis Flasks are manufactured using clean room conditions and have low-protein binding regenerated cellulose membranes to ensure maximum sample recovery and sample purity.

The dialysis flasks are available in distinct colors, corresponding to the pore size (MWCO) of the dialysis membrane: purple (20,000 Daltons), orange (10,000 Daltons), pink (3,500 Daltons), and blue (2,000 Daltons).

Slide-A-Lyzer Dialysis Flasks are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of an inert plastic body. The membrane and flask materials are compatible with most laboratory buffers.

Features of Slide-A-Lyzer Dialysis Flasks:
Easy to use—simply pipette or pour sample into flask and begin dialysis
Fast dialysis—flat flask chamber with two membranes provides high surface area to volume ratio, enabling dialysis of a 250 mL sample in 8 hours to overnight
High recovery—rectangular flask design maximizes recovery of entire sample volume via opening at top of flask
Multiple molecular-weight cut offs—select the membrane MWCO that best suits your sample's molecular weight

Dialysis membrane specifications:
Composition: regenerated-cellulose
Thickness: 0.9–1.2 mil (22.5 to 30 μm); 2K MWCO = 2 mil (50 μm)
Hydration time required before use: 2 minutes
Glycerol content: trace (10K MWCO content is 21%)
Sulfur content: 0.04–0.17%
Heavy metals content: trace

With Slide-A-Lyzer Dialysis Flasks, typical low-molecular weight contaminant removal, buffer exchange, and desalting can be accomplished in eight hours to overnight. The rate of dialysis depends on a variety of factors, including sample volume, size and shape of the molecule being dialyzed, agitation, and temperature. Minimum dialysis time must be determined empirically for each specific system.

TRUSTED_SUSTAINABILITY

Specifications

Description Dialysis Flasks, 2K MWCO
Content And Storage Shipped at ambient temperature. Upon receipt, store at room temperature.
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Flask
MWCO 2.0 kDa
Product Line Slide-A-Lyzer
Quantity 4 Flasks
Sample Volume (Metric) 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.