Learn More
Thermo Scientific™ Slide-A-Lyzer™ G2 Dialysis Cassettes, 10K MWCO, 3 mL, 10 Cassettes PROMO
Description
Slide-A-Lyzer G2 Dialysis Cassettes allow samples to be directly injected with a syringe or pipette through the unique sample port. The cassette design allows air to be removed from inside the cassette to maximize the surface area-to-volume ratio, providing better dialysis performance than dialysis tubing and tube-based systems. The sample is easily recovered through either the pipette or syringe sample ports.
The 10K molecular-weight cutoff (MWCO) cassettes use a low-binding regenerated cellulose that is rated to completely retain proteins and other macromolecules larger than 10,000 Daltons while allowing removal of buffer salts and small contaminants. Membranes and cassettes are manufactured using clean-room conditions to ensure that are contaminant-free.
Features of Slide-A-Lyzer G2 Dialysis Cassettes (10K MWCO):
- Easy-loading—pipette-accessible for easy sample loading and retrieval
- Self-floating—integrated air chambers eliminate the need for float buoys
- Sturdy construction—ensures the highest possible sample integrity and protection
- Superior design—thoroughly researched and tested to provide fast and consistent dialysis with maximum sample recovery
- Versatile—ideal for removing low molecular-weight contaminants, performing buffer exchange and desalting
- Multiple sizes—five cassette capacities to optimally match 0.5 to 70 mL sample volumes
- Gamma-irradiated option—gamma-irradiated cassettes are available for applications requiring conditions that minimize the risk of sample contamination
Gamma-irradiated (sterile) dialysis cassettes
Gamma-irradiated Slide-A-Lyzer G2 Dialysis Cassettes are ideal for researchers culturing cells and microorganisms; purifying viruses, DNA, and RNA; or performing sample preparation for other applications requiring conditions that minimize the risk of sample contamination. The gamma-irradiated cassettes are treated with 20–36 kGy of gamma radiation, a dosage that has been determined to sterilize the units based on both trypticase soy broth medium for identifying total growth and fluid thioglycollate medium for identifying anaerobic growth. However, a confirmation of sterility is not provided for individual lots, as no further sterility testing is conducted post irradiation.
Membrane specifications:
- Composition: regenerated cellulose synthesized by the viscose method
- Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
- Hydration time required before use: 2 minutes
- Glycerol content: 21%
- Sulfur content: 0.1–0.15%
- Heavy metals content: trace
Volume ranges:
- 0.5 mL = 0.25 to 0.75 mL
- 3 mL = 1 to 3 mL
- 15 mL = 5 to 15 mL
- 30 mL = 10 to 30 mL
- 70 mL = 25 to 70 mL
Specifications
Specifications
Content And Storage | Store at room temperature |
Purification Target | Buffer Exchange, Protein |
Membrane | Cellulose Membrane, Dialysis |
Format | Cassette |
MWCO | 10 kDa |
Product Line | Slide-A-Lyzer™ |
Quantity | 10 Cassettes |
Sample Volume (Metric) | 3 mL |
Product Type | G2 Dialysis Cassettes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.