Learn More
Thermo Scientific™ Slide-A-Lyzer™ G2 Dialysis Cassettes, 2K MWCO, 70 mL, 6 Cassettes PROMO
Description
Slide-A-Lyzer G2 Dialysis Cassettes allow samples to be directly injected with a syringe or pipette through the unique sample port. The cassette design allows air to be removed from inside the cassette to maximize the surface area-to-volume ratio, providing better dialysis performance than dialysis tubing and tube-based systems. The sample is easily recovered through either the pipette or syringe sample ports.
Features of Slide-A-Lyzer G2 Dialysis Cassettes (2K MWCO):
- Easy-loading—pipette-accessible for easy sample loading and retrieval
- Self-floating—integrated air chambers eliminate the need for float buoys
- Sturdy construction—ensures the highest possible sample integrity and protection
- Superior design—thoroughly researched and tested to provide fast and consistent dialysis with maximum sample recovery
- Multiple sizes—five cassette capacities to optimally match 0.5 to 70 mL sample volumes
- Versatile—ideal for removing low-molecular weight contaminants, performing buffer exchange, and desalting
Volume ranges:
- 0.5 mL = 0.25 to 0.75 mL
- 3 mL = 1 to 3 mL
- 15 mL = 5 to 15 mL
- 30 mL = 10 to 30 mL
- 70 mL = 25 to 70 mL
Specifications
Specifications
Content And Storage | Store at room temperature |
Purification Target | Buffer Exchange, Protein |
Membrane | Cellulose Membrane, Dialysis |
Format | Cassette |
MWCO | 2.0 kDa |
Product Line | Slide-A-Lyzer™ |
Quantity | 6 Cassettes |
Sample Volume (Metric) | 70 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.