Learn More
United Scientific™ Slide Tray, Cardboard, 5 Pk.
SDP
Catalog No. 1001309 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Slide Capacity:
20
30
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Slide Capacity
10-013-09 20
10-013-10 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-013-09 Supplier United Scientific™ Supplier No. 62603PK5
Add to Cart
Add to Cart
Protects your slides while holding and transferring them.
Designed as a convenient solution for holding and transferring microscope slides. Fold-over cover helps protect the slides. The flat surface area on the inside of the cover is ideal for labeling or writing any necessary information. Recessed compartments protect the slides when the cover is closed. Thumb cuts on the sides allow you to easily remove each slide. Accommodates microscope slides in two sizes: 75.0 mm L x 25.0 mm W x 0.8 - 1.2 mm D and 76.0 mm L x 26.0 mm W x 0.8 - 1.2 mm D
  • Recessed compartments to protect slides when closed
  • Heavy duty cardboard
  • Thumb cuts for easy removal of slides
  • Flat writing surface for specimen identification
Specifications
Type Slide Storage Tray
Slide Capacity 20
Autoclavable Not Autoclavable