Description
- Assorted color clasps are ideal for color coding identification and to keep slide folder flaps securely closed
- Clasps can be labeled with a permanent marker or a label can be attached
Specifications
Specifications
| Type | Slide Storage System |
| Color | Assorted |
| Device Holds | Cardboard Slide Trays |
| Dimensions (L x W x H) | 2.2 x 1.7 x 0.6 in. |