Learn More
Epredia™ SlideMate™ Laser Printer Filters
GSA_VA
Catalog No. 4000850 Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment): SlideMate™ Laser Printer
Catalog No.
40-008-50 For Use With (Equipment): SlideMate™ Laser Printer
1 options
This item is not returnable. View return policy
Catalog No. 40-008-50 Supplier Epredia™ Supplier No. 4000850
Add to Cart
Add to Cart
This item is not returnable. View return policy
SlideMate™ Laser Printer filters (5 per pack) for use with the SlideMate Laser printer
  • Integrate SlideMate laser into any laboratory workflow
  • True on-demand slide barcode printing
  • Filters for use with the SlideMate Laser printer
  • Mounted on the rear of the instrument for additional filtration of particles from laser slide printing technology
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Filter
For Use With (Equipment) SlideMate™ Laser Printer
Includes One Pk. of 5 filters