Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ SlideMate™ Adhesion Microscope Slides
Shop All Epredia Products
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Description
- Manufactured in the USA; SlideMate slides are smooth, three-quarter inch white, blue, green, pink or yellow tab slides validated to work with the SlideMate AS slide printer.
- The tab is impervious to virtually all common laboratory chemicals and reagents.
- Available in adhesion and non-adhesion formulas. SlideMate Plus Adhesion Slides feature adhesion coating for routine clinical applications.
- Epredia SlideMate slides are made from extra-white glass, manufactured to high quality standards. The clarity achieved during the manufacturing process helps ensure high levels of transparency to aid in accurate diagnoses.
- SlideMate slides come pre-cleaned, flat with finely ground edges. Each package comes wrapped to help protect against humidity and moisture. The packaging is also designed to assist with loading the slides into SlideMate printer hoppers to prevent contamination due to handling.
Specifications
Specifications
Length (Metric) | 75 mm |
Width (Metric) | 25 mm |
Thickness (Metric) | 1 mm |
No. per Case | 1440/Cs. |
No. per Pack | 144 Pk. |
Material | Glass |
For Use With (Equipment) | SlideMate Printers |
Edges | Ground |
Color | Yellow |
Coating | Adhesion |
Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction