Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SLIGRL-NH2
GREENER_CHOICE

Catalog No. 14681r
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

PAR2-activating peptide

SLIGRL-NH2 is an agonist peptide derived from the N-terminus of protease-activated receptor-2 (PAR2). Activates PAR2 (EC50 ∽ 5 μM) and facilitates gastrointestinal transit in mice in vivo.

Control Peptide also available.

Specifications

Purity 0.95
Content And Storage Store at -20°C
Description SLIGRL-NH2
Molecular Weight (g/mol) 656.82
Product Type SLIGRL-NH2
Quantity 1 mg
Sequence Ser-Leu-Ile-Gly-Arg-Leu-NH2

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.