Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Micronova™ SlimLine™ Buckets with Casters
Shop All Micronova Manufacturing Products

Click to view available options
Color:
Dark Blue
Yellow
Description
For use with SlimLine™ Cleanroom Cart System
- For use with SlimLine or SlimT Cleanroom Cart System
- 6 gallon capacity
- Autoclavable
- Available in blue, red, green or yellow
Specifications
Specifications
| Product Type | SlimLine Bucket |
| Material | Polypropylene Bucket, Nickel-plated Caster, Thermoplastic Rubber Wheel |
| Autoclavable | Autoclavable |
| Care Instructions | Store at Ambient Temperature. To Autoclave - do not stack - invert Bucket to allow drainage. |
| Color | Dark Blue |
| For Use With (Application) | Cleaning Walls, Floors and Ceilings |
| For Use With (Equipment) | SlimLine or SlimT Cleanroom Cart System |
| Sterility | Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction