Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ SlimLine™ Cleanroom Cart System
SDP

Catalog No. p-3350298
Click to view available options
Product Type:
Double Bucket Cart
Triple Bucket Cart
Color:
Blue, Yellow
Blue, Yellow, Red

Includes

Each system supplied with 6-gal. (22.7L) polypropylene buckets. Two-bucket system includes one blue and one yellow bucket; three-bucket system includes one blue, one red, and one yellow bucket. Optional wringer, basket, bucket liner and replacement buckets sold separately.

A durable, double or triple bucket cart system with an electropolished, stainless steel frame, optional wringer bar, and three grip-it holders for easy storage and interchange of handles and mop heads. Systems comes standard with color coded, 22.7 liter (6 gal.) polypropylene buckets.

The Micronova SlimLine™ Stainless Steel Cart Systems include an electropolished stainless steel cart and come standard with either two or three, color-coded, polypropylene buckets to distinguish between clean and rinse buckets. The three grip-it holders at the back of the cart allow for easy interchange and storage of handles and mop heads. An optional wringer bar accommodates either the Downpress or Flat Head Wringers, allowing placement above the rinse bucket for simplified wringing.

  • Cart is crafted from electropolished stainless steel for durability and long life
  • Includes three grip-it holders at the back of the cart
  • Easy storage and interchange of handles and mop heads
  • Comes standard with color-coded, 6-gallon (22-L), polypropylene buckets
  • Double bucket system comes with 1 blue and 1 yellow bucket
  • Triple bucket system includes an additional red bucket
  • Optional wringer bar allows wringers to be placed above the rinse bucket
  • Buckets are best autoclaved in the inverted position
  • Entire unit is autoclavable

Specifications

Autoclavable Autoclavable
Product Type Triple Bucket Cart
Capacity (English) 18 gal. (6 gal./Bucket)
Capacity (Metric) 68.1 L (22.7 L/bucket)
Color Blue, Yellow, Red
For Use With (Equipment) SlimLine™ Cleanroom Cart System
Includes 3 Buckets
Material Stainless Steel
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.