Learn More
Simport™ Scientific Slimsette™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks
Catalog No. 22038550 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-550 Gray
22-038-543 White
22-038-544 Pink
22-038-545 Green
22-038-546 Yellow
22-038-547 Blue
22-038-548 Peach
22-038-549 Tan
22-038-553 Lilac
22-038-554 Orange
22-038-555 Aqua
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 22-038-550 Supplier Simport™ Scientific Supplier No. M5109T
Add to Cart
Add to Cart
This item is not returnable. View return policy
Secure specimens during processing with Slimsette™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks.
  • In Quickload™ stacks
  • 45° Anterior writing area
  • Recessed cover
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Quantity 2000/Pk.
Color Gray
Material Acetal